Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel™ QuickVue™ TLI H. pylori Stool Antigen Test
SDP

Catalog No. 23043040 Shop All QuidelOrtho Products
Change view
Click to view available options
Quantity:
25/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-043-040 25/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-043-040 Supplier Quidel™ Supplier No. 20343

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A rapid membrane enzyme immunoassay for the qualitative detection of Helicobacter pylori specific antigen in a single use cassette

  • Results in 30 minutes
  • High sensitivity and specificity in determining active infection before and after eradication therapy
  • Can be used with various sample types, including fresh, frozen, Cary Blair, or C&S samples
  • Samples can be stored refrigerated or at room temperature for 96 hours
  • It uses human fecal specimens to aid in the diagnosis of H. pylori infection and to demonstrate loss of H. pylori antigen following treatment
  • The test can be used with unpreserved fecal specimens and fecal specimens preserved in transport media from patients suspected of H. pylori infection

Specifications

Content And Storage 2°C to 8°C
Clia Complexity Moderate
Sample Type Human Fecal
Sensitivity 97%
Includes Internal and External Controls
Quantity 25/Kit
Test Time 30 min. or less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.