Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel AmpliVue™ Bordetella Assay Kits
SDP

Catalog No. 23043138 Shop All QuidelOrtho Products
Change view
Click to view available options
Type:
Control
Starter Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
23-043-138 Control
23-043-139 Starter Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-043-138 Supplier Quidel Supplier No. M117
Add to Cart
Add to Cart
This item is not returnable. View return policy

In vitro diagnostic test for the qualitative detection of Bordetella pertussis nucleic acids

  • Small, self-contained hand-held cassette
  • Convenient, easy-to-use, disposable device
  • HDA technology; Rapid method of nucleic acid amplification that does not require thermal cycler
  • Lyophilized reagents; Easy-to-use format, just rehydrate and add sample
  • Reagents are stable at higher temperatures than liquid reagents; Small easy-to-use heat blocks
  • Eliminates need for dedicated molecular space and costly capital equipment; Allows for testing in smaller labs
  • CLIA moderately complex
  • Requires minimal training
  • Refrigerated storage; No freezer needed; 2°C to 8°C storage
  • Long shelf life; 2 years from date of manufacture
  • Room temperature set up; No ice or cooling block required

Specifications

Content And Storage Controls storage conditions: 2°C to 8°C
Sample Type Nasopharyngeal Swab
Type Control
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.