Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel Molecular Strep A+G Control Set
GSA_VASDP

Catalog No. 23043150 Shop All QuidelOrtho Products
Change view
Click to view available options
Includes:
Control Kit (2 vials)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
23-043-150 Control Kit (2 vials)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-043-150 Supplier Quidel Supplier No. M111
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Unassayed Control Set for in vitro diagnostic use. Positive and Negative.

The Quidel MolecularStrep A+G Control Set is intended to be used as full process controls in molecular testing.  This unassayed Control Set for in vitro diagnostic use consists of a highly purified, inactivated mix of bacteria which includes Group A Streptococcus and Group G Streptococcus.The negative control is Group A Streptococcus and Group G Streptococcus free matrix.

  • Contains positive and negitive controls
  • Serves as an external processing and extraction
  • For use with  Solana™ GAS, Solana Step Complete, and Lyra Direct Strep

 

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Content And Storage 2°C to 8°C
For Use With (Application)  Solana™ GAS (25μL), Solana Step Complete (25μL) and Lyra Direct Strep (25μL).
Includes Control Kit (2 vials)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.