Learn More
Quidel Corporation MicroVue C4a Enzyme Immunoassay 96 tests
SDP
Supplier:  Quidel Corporation A035
Encompass
The MicroVue C4a Enzyme Immunoassay is for the measurement of C4a in human or primate serum, plasma, and other biological or experimental samples.
Catalog No. NC0568629