Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel Quidel Corporation InflammaDry Control Set
SDP

Catalog No. 23043054 Shop All QuidelOrtho Products
Change view
Click to view available options
Quantity:
1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-043-054 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-043-054 Supplier Quidel Supplier No. RPSDESTD
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Quidel Corporation InflammaDry Control Set contains 1 positive, 1 negative, and a diluent. Lypholized

Specifications

Content And Storage Room temperature, not to exceed 86°F/30°C
CE Marker Yes
Detectable Analytes MMP-9
For Use With (Application) InflammaDry test kit
Includes Lyophilized positive, lyophilized negative, diluent
Packaging Type 1/Ea.
Quantity 1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.