Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel Quidel Corporation InflammaDry Test Kit
SDP

Catalog No. 23043050 Shop All QuidelOrtho Products
Change view
Click to view available options
Quantity:
20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-043-050 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-043-050 Supplier Quidel Supplier No. RPSID20U
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Quidel Corporation InflammaDry Test Kit is a rapid, in-office, CLIA-waived test that detects elevated levels of MMP-9, an inflammatory marker elevated in tears of patients with dry-eye

  • Accurately identifies patients with dry eye in 4 steps, allowing for optimal treatment methods and better quality of life
  • Accurate: 81% sensitivity, 98% specificity
  • Easy to use: 4 simple steps, can be performed by a technician
  • Cost effective: single use, disposable test
  • Fast: results in as soon as 10 minutes

Specifications

Content And Storage 39°F to 77°F
CE Marker Yes
Detectable Analytes MMP-9
For Use With (Application) Dry Eye
Includes Individually Packaged Sample Collector (20), Individually Packaged Test Cassettes (20), Buffer Vial (20): buffered salt solution containing 0.1% Sodium Azide, as a preservative, Package Insert (1)
Packaging Type 20 Test Kit
Quantity 20
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.