Description
Flocked swabs provide improved sample collection. The velvet brush-like texture rapidly and efficiently dislodges cells and collects liquid by capillarity. With no internal mattress core to disperse and entrap the precious sample like traditional fiber wound swabs, the entire sample stays close to the surface for fast and complete elution.
- Nylon flocked, dry swabs in peel pouch
- Nylon tip; plastic applicator; sterile; individually wrapped
- Perpendicular nylon fibers act like a soft brush and allow improved collection of cell samples
- Capillary action between the nylon fiber strands facilitates strong hydraulic uptake of liquid samples
- Ergonomic and anatomic design for improved patient comfort and efficiency in specimen collection
- Rapid automatic elution: instant and spontaneous release into liquid media
- Quantitative volume transfer: measurable and consistent uptake and transfer from patient to test tube
- Significantly improve quantity of sample collected and released into various culture and assay systems, improving quality of diagnostics
- Flocked swabs have been shown to improve the sensitivity of various diagnostic tests
- Certified DNase, RNase-free and human DNA-free, making them ideal for collection and transfer of minute amounts of DNA
- Breakpoint distance: 80mm
- Packaging: 10 x 100 pcs
Specifications
Specifications
| Product Type | Minitip Flocked Swab |
| For Use With (Equipment) | For use with QuickVue RSV, QuickVue RSV 10 and QuickVue Influenza A+B tests |
| Quantity | 100/Pk. |
| Tip Material | Nylon Flock |