Learn More
Quidel Molecular C. difficile Control Set
SDP
Catalog No. 23043141 Shop All QuidelOrtho Products
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
23-043-141 Each
1 options
This item is not returnable. View return policy
Catalog No. 23-043-141 Supplier Quidel Supplier No. M108
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Includes
C. difficile Positive Control (M5040) and Negative Control (M503) : 2mL each
To be used as full process controls in molecular testing
  • Unassayed set, consists of highly purified, inactivated strain of C. difficile (NAP1) and a C. difficile free matrix
Certifications
CE marked
Specifications
Type C. difficile Control Set
Includes C. difficile Positive Control and Negative Control of 2mL each
Quantity Each