Learn More
Quidel Molecular GBS Control Set
SDP
Catalog No. 23043143 Shop All QuidelOrtho Products
Change view
Click to view available options
Quantity:
20 Reactions
Catalog No. Quantity
23-043-143 20 Reactions
1 options
This item is not returnable. View return policy
Catalog No. 23-043-143 Supplier Quidel Supplier No. M116
Add to Cart
Add to Cart
This item is not returnable. View return policy
To be used as full process controls in molecular testing
  • Unassayed external set includes 1mL positive and 2mL negative control
  • Number of reactions per kit: 20
Specifications
Type Molecular GBS Control Set
Includes Positive Control and Negative Control
Quantity 20 Reactions