Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel QuickVue™ Chlamydia Test
SDP

Catalog No. 23038170 Shop All QuidelOrtho Products
Change view
Click to view available options
Type:
Chlamydia Swab
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
23-038-170 Chlamydia Swab
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-038-170 Supplier Quidel Supplier No. 20228

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Materials for 25 tests with external positive and negative controls

Provides accurate and easy-to-interpret results in 12 minutes or less

  • CLIA Complexity: Non-Waived
  • CPT Code: 87810
  • Sensitivity: 92% (Overall)
  • Specificity: 99% (Overall)
  • Lateral-flow immunoassay for the qualitative detection of Chlamydia trachomatis antigen directly from endocervical swab or cytology brush specimens
  • Simple extraction procedure requires the addition of only two reagents
  • Two-color endpoint for easy-to-read results
  • Highly accurate in asymptomatic patient population
  • Built-in controls confirm test has been performed correctly
  • For in vitro diagnostic use

Specifications

Content And Storage Room Temperature
Type Chlamydia Swab
Quantity 25 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.