Learn More
Quidel QuickVue™ H. pylori Test
Shop All QuidelOrtho Products
Click to view available options
Quantity:
10 Tests
30 Tests
Description
Includes
Each complete kit contains testing cassettes, positive and negative controls, transfer pipets and capillary tubes, package insert and procedure card
- CLIA Complexity: Waived (whole blood); Non-waived (serum and plasma)
- CPT Codes: 86318 (serum/plasma), 86318QW (whole blood)
- Lateral-flow immuno-chromatographic assay
- Test results in five minutes; 92% overall agreement with EIA
- Tests serum and plasma
- Two-color endpoint for easy-to-read and interpret results
- Simple one-step procedure from only two hanging drops of blood; serum or plasma can also be used
- For in vitro diagnostic use
Specifications
Specifications
| Content And Storage | Room Temperature |
| Clia Complexity | Waived, Non-waived |
| Detectable Analytes | H. pylori IgG |
| Type | H. pylori Test Kit |
| Includes | Testing cassettes, positive and negative control, transfer pipets and capillary tubes, package insert and procedure card, positive and negative external control |
| Quantity | 30 Tests |