Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel QuickVue™ iFOB Test
SDP

Catalog No. p-3507624 Shop All QuidelOrtho Products
Change view
Click to view available options
Quantity:
1 Case
12 Case
6 Case
8 Case
Type:
iFOB Control Set
iFOB Spec Collect, Return Kit
iFOB Specimen Collection Kit
iFOB Test Kit
iFOBT Cassette Kit
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
23-038-064 12 Case iFOB Spec Collect, Return Kit
23-038-063 6 Case iFOB Test Kit
23-038-065 1 Case iFOB Control Set
23-038-066 8 Case iFOB Test Kit
23-038-067 6 Case iFOB Specimen Collection Kit
23-038-068 6 Case iFOBT Cassette Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 23-038-064 Supplier Quidel Supplier No. 20196
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Individually wrapped test cassettes, package insert, procedure card, specimen collection and return kits, each consisting of: Specimen collection tube, with 2mL FOB buffer (1 x PBS with 0.02% sodium azide) and patient identification label, specimen collection paper with adhesive, specimen pouch, absorbent sleeve, return mailer, patient instructions

Immunochemical FOB test—a more analytically sensitive alternative to Guiac methods

  • CLIA Complexity: Waived
  • Specificity: Specific to human hemoglobin
  • Analytical Sensitivity: 50ng hHb/mL buffer or 50μg hHb/g feces
  • POL agreement: 98%
  • Results in 5 to 10 minutes; positive results may become visible sooner
  • Patient friendly, easy to use
  • Patients are not required to adhere to strict dietary restrictions

Order Info

  • Additional specimen collection and return kits available separately in convenient 10-kit size
  • External controls available separately

Specifications

Clia Complexity Waived
Type iFOB Spec Collect, Return Kit
For Use With (Application) To detect the presence of Blood in stool specimens
Quantity 12 Case
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.