Learn More
Quidel QuickVue™ Influenza Swabs
SDP
Catalog No. 23038172 Shop All QuidelOrtho Products
Change view
Click to view available options
Quantity:
25 Swabs
Catalog No. Quantity
23-038-172 25 Swabs
1 options
This item is not returnable. View return policy
Catalog No. 23-038-172 Supplier Quidel Supplier No. 20103
Add to Cart
Add to Cart
This item is not returnable. View return policy
Nasal swab pack. 25 individually pouched sterile foam swabs. For use with QuickVue Influenza, QuickVue Influenza A+B and Sofia Influenza A+B FIA tests only.
See also:  Quidel QuickVue™ Influenza Test Kit (23-043-044).
Specifications
Detectable Analytes Influenza A and B
Type Influenza Swab
Quantity 25 Swabs