Learn More
Description
Sample directly from nasopharyageal swab or nasopharyngeal aspirate/wash specimens for symptomatic pediatric patients (five years of age and younger). A single sample can be used to run both the QuickVue RSV 10 test and the QuickVue Influenza A+B test.
Test results are available in 10 minutes or less.
Features and Benefits
- Dipstick test format is easy to use; approximately 30 sec. hands-on time
- Two-color result provides easy to read and interpret results
- 10 min. to result allows prompt diagnosis and appropriate patient management while patient is still in office or hospital
- Only three steps and one reagent
- No reagent warm-up; room temperature storage, use as needed without waiting
- Built-in internal controls provide verification of test strip functional integrity, which increases confidence in results
- Kit includes external controls in order to facilitate internal laboratory quality control
- All components needed to perform 25 tests are contained in the kit, including nasopharyngeal swabs
Specifications
Specifications
| Detectable Analytes | Respiratory Syncytial Virus |
| Format | Dipstick |
| Sample Type | Nasal Aspirate, Nasal Swab, Nasal Wash |
| Type | RSV 10 Test Kit |
| Includes | Nasopharyngeal swabs, external control and built-in internal control |
| Quantity | 25 Tests |
| Test Time | ≤10 min. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
