Learn More
Quidel QuickVue™ Strep A Control Swab Set
SDP
Catalog No. 23043046 Shop All QuidelOrtho Products
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
23-043046 Each
1 options
This item is not returnable. View return policy
Catalog No. 23-043046 Supplier Quidel Supplier No. 00345
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
For use with Quidel QuickVue Flex Strep A and QuickVue In-Line™ One-Step Strep A Tests
Specifications
Detectable Analytes Streptococci (Group A)
Type Positive and negative Control Swab Set
Includes 6 Positive and 6 Negative control swabs
Quantity Each