Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel Serum hCG Control Set
GSA_VASDP

Catalog No. 120115 Shop All QuidelOrtho Products
Change view
Click to view available options
Quantity:
3 x 5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-011-5 3 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 12-011-5 Supplier Quidel Supplier No. 00281
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with Quidel CARDS Q.S. hCG and QuickVue™ Semi-Q hCG Combo Tests

  • 3 x 5mL

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Content And Storage Refrigerated
Type Serum hCG Control Set
Includes hCG testing, pregnancy tests, Serum hCG control set
Intended Use Quidel CARDS Q.S. hCG and QuickVue Semi-Q hCG combo tests, provide an aid in the interpretation of positive and negative test results and verify proper test performance
Quantity 3 x 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.