Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel Sofia™ Influenza A+B FIA Kit
GSA_VASDP

Catalog No. 23043070 Shop All QuidelOrtho Products
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-043-070 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-043-070 Supplier Quidel Supplier No. 20218
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For rapid differential diagnosis of acute influenza A and influenza B viral infections

  • CLIA Complexity: Waived for Nasal and Nasopharyngeal swab specimens
  • CPT codes: 87804/87804-59
  • Detects and differentiates influenza A and B viral infections
  • For use with Sofia Analyzer
  • Results in 15 minutes
  • Enhanced assay sensitivity
  • In vitro diagnostic use only
  • Employs immunofluorescence
  • Detect influenza nucleoprotein antigens in nasal swab and nasopharyngeal swab and NPA wash samples from symptomatic patients

Order Info

25-test kit

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Detectable Analytes Influenza A, Influenza B
Sample Type Nasal Swab, Nasopharyngeal Swab, Nasopharyngeal Wash/Aspirate
Type Influenza A and B FIA Kit
Quantity Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.