Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel Sofia™ Strep A FIA CLIA-Waived
SDP

Catalog No. 23043083 Shop All QuidelOrtho Products
Change view
Click to view available options
Quantity:
25 Test Kit
Type:
Streptococcus A FIA
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
23-043-083 25 Test Kit Streptococcus A FIA
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-043-083 Supplier Quidel Supplier No. 20274
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

25 Test-Kit

  • Individually Packaged Cassettes (25): Polyclonal rabbit anti-Group A Streptococcus antibodies
  • Reagent Tubes (25)
  • Reagent Solution Bottles (25): 4M Sodium Nitrite and 0.2M Acetic Acid inside glass ampoule
  • Fixed Volume Pipettes (25)
  • Sterile Rayon Throat Swabs (25)
  • Positive Control Swab (1): Swab is coated with heat-inactivated, non-infectious Group A Streptococcus
  • Negative Control Swab (1): Swab is coated with heat-inactivated, non-infectious Group C Streptococcus
  • Package Insert (1)
  • Quick Reference Instructions (1)
  • QC Card (located on kit box)
  • Printer Paper (1)

For rapid qualitative detection of Group A Streptococcal antigens from throat swabs of symptomatic patients

The Sofia Strep A+ FIA employs immunofluorescence technology that is used with the Sofia Analyzer to detect Group A Streptococcal antigen.

  • For use with Sofia Analyzer
  • CLIA-Waived
  • Highly sensitive and specific with enhanced assay accuracy
  • Analyzes and provides objective result versus visually read tests
  • Involves antigenic components extraction of Group A Streptococcus (GAS) bacteria
  • Results in 5 minutes

Applications:

Healthcare industry for diagnostic purposes

Specifications

Content And Storage Room Temperature
Clia Complexity Waived
Detectable Analytes Streptococci (Group A) Antigen
Sample Type Throat Swab
Type Streptococcus A FIA
Includes Cassettes, reagent tubes, reagent solution (contains Sodium Nitrite and Acetic Acid), reagent dropper tips, sterile rayon throat swabs, positive control swab and negative control swab
Quantity 25 Test Kit
Test Time 5 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.