Learn More
Medchemexpress LLC HY-50714 5mg Medchemexpress, Quiflapon (sodium) CAS:147030-01-1 Purity:>98%
SDP
Catalog No. 502082252
Encompass_Preferred
Medchemexpress, HY-50714 5mg Quiflapon (sodium) CAS:147030-01-1 Quiflapon sodium (MK-591 sodium) is a selective and specific 5-Lipoxygenase-activating protein (FLAP) inhibitor. Quiflapon sodium is an orally active Leukotriene biosynthesis inhibitor. Induces apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2252 Supplier Medchemexpress LLC Supplier No. HY507145MG
Medchemexpress, HY-50714 5mg Quiflapon (sodium) CAS:147030-01-1 Quiflapon sodium (MK-591 sodium) is a selective and specific 5-Lipoxygenase-activating protein (FLAP) inhibitor. Quiflapon sodium is an orally active Leukotriene biosynthesis inhibitor. Induces apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.