Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Quik-Align™ No Water Picto Polyester Label

Catalog No. 17200440 Shop All Brady Corporation Products
Change view
Click to view available options
Height (English):
4 in.
Width (English):
4.25 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (English) Width (English)
17-200-440 4 in. 4.25 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-200-440 Supplier Brady™ Supplier No. 175334
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Easily apply outdoor-durable Quik-Align™ labels to identify sites, chemical containers or other points of emphasis where unusual reactivity with water could occur.

  • Card-mounted polyester film characters feature pressure-sensitive adhesive
  • Flexible material conforms to most surfaces and holds up to harsh environments, abrasion, and repeated spills and cleanings
  • Weathers up to 8 years outdoors, even longer indoors, and withstands wide temperature variations (-40°F to +230°F)

Specifications

Material Laminated Polyester
Quantity 10 Labels/Pk.
Adhesive Type Permanent Acrylic
For Use With (Application) Facility Identification, Hazard Communication Identification, Safety Identification
Height (English) 4 in.
Width (English) 4.25 in.
Width (Metric) 107.95 mm
Temperature Range (Metric) -40°C to +110°C
Temperature Range (English) -40°F to +230°F
Displayed Text W
Color Black on White
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.