Learn More
Agilent Technologies QUIK CHANGE MULTI 10 REACTIONS
Catalog No. NC9364692
Encompass
Kit, Multi Site Directed Mutagenesis; QuikChange;; Multi-site mutagenesis in a single reac.; Contains DNA polymerase blend, template digestion enzyme, competent cells; 1/KT (30 rxn)
Catalog No. NC9364692 Supplier Agilent Technologies Supplier No. 200531
Kit, Multi Site Directed Mutagenesis; QuikChange;; Multi-site mutagenesis in a single reac.; Contains DNA polymerase blend, template digestion enzyme, competent cells; 1/KT (30 rxn)