Learn More
Hampton Research QUIK SCREEN NOA1 185ML
SDP
Catalog No. 501800
Encompass
HR2-921-A1/Quik Screen #A1/Grid Screen Salt HT (E01), 185 ml - 0.8 M Sodium phosphate monobasic monohydrate / Potassium phosphate dibasic pH 5.0. Please note items from this supplier are non-returnable.
Catalog No. 50-180-0 Supplier Hampton Research Supplier No. HR2921A1
HR2-921-A1/Quik Screen #A1/Grid Screen Salt HT (E01), 185 ml - 0.8 M Sodium phosphate monobasic monohydrate / Potassium phosphate dibasic pH 5.0. Please note items from this supplier are non-returnable.