Learn More
Hampton Research QUIK SCREEN NOA5 185ML
SDP
Catalog No. 501804
Encompass
HR2-921-A5/Quik Screen #A5/Grid Screen Salt HT (E05), 185 ml - 0.8 M Sodium phosphate monobasic monohydrate / Potassium phosphate dibasic pH 7.5. Please note items from this supplier are non-returnable.
Catalog No. 50-180-4 Supplier Hampton Research Supplier No. HR2921A5
HR2-921-A5/Quik Screen #A5/Grid Screen Salt HT (E05), 185 ml - 0.8 M Sodium phosphate monobasic monohydrate / Potassium phosphate dibasic pH 7.5. Please note items from this supplier are non-returnable.