Learn More
Hampton Research QUIK SCREEN NOA6 185ML
SDP
Catalog No. 501805
Encompass
HR2-921-A6/Quik Screen #A6/Grid Screen Salt HT (E06), 185 ml - 0.8 M Sodium phosphate monobasic monohydrate / Potassium phosphate dibasic pH 8.2. Please note items from this supplier are non-returnable.
Catalog No. 50-180-5 Supplier Hampton Research Supplier No. HR2921A6
HR2-921-A6/Quik Screen #A6/Grid Screen Salt HT (E06), 185 ml - 0.8 M Sodium phosphate monobasic monohydrate / Potassium phosphate dibasic pH 8.2. Please note items from this supplier are non-returnable.