Learn More
Hampton Research QUIK SCREEN NOB1 185ML
SDP
Catalog No. 501806
Encompass
HR2-921-B1/Quik Screen #B1/Grid Screen Salt HT (E07), 185 ml - 1.0 M Sodium phosphate monobasic monohydrate / Potassium phosphate dibasic pH 5.0. Please note items from this supplier are non-returnable.
Catalog No. 50-180-6 Supplier Hampton Research Supplier No. HR2921B1
HR2-921-B1/Quik Screen #B1/Grid Screen Salt HT (E07), 185 ml - 1.0 M Sodium phosphate monobasic monohydrate / Potassium phosphate dibasic pH 5.0. Please note items from this supplier are non-returnable.