Learn More
Hampton Research QUIK SCREEN NOB2 185ML
SDP
Catalog No. 501807
Encompass
HR2-921-B2/Quik Screen #B2/Grid Screen Salt HT (E08), 185 ml - 1.0 M Sodium phosphate monobasic monohydrate / Potassium phosphate dibasic pH 5.6. Please note items from this supplier are non-returnable.
Catalog No. 50-180-7 Supplier Hampton Research Supplier No. HR2921B2
HR2-921-B2/Quik Screen #B2/Grid Screen Salt HT (E08), 185 ml - 1.0 M Sodium phosphate monobasic monohydrate / Potassium phosphate dibasic pH 5.6. Please note items from this supplier are non-returnable.