Learn More
Hampton Research QUIK SCREEN NOB3 185ML
SDP
Catalog No. 501808
Encompass
HR2-921-B3/Quik Screen #B3/Grid Screen Salt HT (E09), 185 ml - 1.0 M Sodium phosphate monobasic monohydrate / Potassium phosphate dibasic pH 6.3. Please note items from this supplier are non-returnable.
Catalog No. 50-180-8 Supplier Hampton Research Supplier No. HR2921B3
HR2-921-B3/Quik Screen #B3/Grid Screen Salt HT (E09), 185 ml - 1.0 M Sodium phosphate monobasic monohydrate / Potassium phosphate dibasic pH 6.3. Please note items from this supplier are non-returnable.