Learn More
Hampton Research QUIK SCREEN NOB4 185ML
SDP
Catalog No. 501809
Encompass
HR2-921-B4/Quik Screen #B4/Grid Screen Salt HT (E10), 185 ml - 1.0 M Sodium phosphate monobasic monohydrate / Potassium phosphate dibasic pH 6.9. Please note items from this supplier are non-returnable.
Catalog No. 50-180-9 Supplier Hampton Research Supplier No. HR2921B4
HR2-921-B4/Quik Screen #B4/Grid Screen Salt HT (E10), 185 ml - 1.0 M Sodium phosphate monobasic monohydrate / Potassium phosphate dibasic pH 6.9. Please note items from this supplier are non-returnable.