Learn More
Hampton Research QUIK SCREEN NOB5 185ML
SDP
Catalog No. 501810
Encompass
HR2-921-B5/Quik Screen #B5/Grid Screen Salt HT (E11), 185 ml - 1.0 M Sodium phosphate monobasic monohydrate / Potassium phosphate dibasic pH 7.5. Please note items from this supplier are non-returnable.
Catalog No. 50-181-0 Supplier Hampton Research Supplier No. HR2921B5
HR2-921-B5/Quik Screen #B5/Grid Screen Salt HT (E11), 185 ml - 1.0 M Sodium phosphate monobasic monohydrate / Potassium phosphate dibasic pH 7.5. Please note items from this supplier are non-returnable.