Learn More
Hampton Research QUIK SCREEN NOC2 185ML
SDP
Catalog No. 501813
Encompass
HR2-921-C2/Quik Screen #C2/Grid Screen Salt HT (F02), 185 ml - 1.4 M Sodium phosphate monobasic monohydrate / Potassium phosphate dibasic pH 5.6. Please note items from this supplier are non-returnable.
Catalog No. 50-181-3 Supplier Hampton Research Supplier No. HR2921C2
HR2-921-C2/Quik Screen #C2/Grid Screen Salt HT (F02), 185 ml - 1.4 M Sodium phosphate monobasic monohydrate / Potassium phosphate dibasic pH 5.6. Please note items from this supplier are non-returnable.