Learn More
Hampton Research QUIK SCREEN NOC3 185ML
SDP
Catalog No. 501814
Encompass
HR2-921-C3/Quik Screen #C3/Grid Screen Salt HT (F03), 185 ml - 1.4 M Sodium phosphate monobasic monohydrate / Potassium phosphate dibasic pH 6.3. Please note items from this supplier are non-returnable.
Catalog No. 50-181-4 Supplier Hampton Research Supplier No. HR2921C3
HR2-921-C3/Quik Screen #C3/Grid Screen Salt HT (F03), 185 ml - 1.4 M Sodium phosphate monobasic monohydrate / Potassium phosphate dibasic pH 6.3. Please note items from this supplier are non-returnable.