Learn More
Hampton Research QUIK SCREEN NOC4 185ML
SDP
Catalog No. 501815
Encompass
HR2-921-C4/Quik Screen #C4/Grid Screen Salt HT (F04), 185 ml - 1.4 M Sodium phosphate monobasic monohydrate / Potassium phosphate dibasic pH 6.9. Please note items from this supplier are non-returnable.
Catalog No. 50-181-5 Supplier Hampton Research Supplier No. HR2921C4
HR2-921-C4/Quik Screen #C4/Grid Screen Salt HT (F04), 185 ml - 1.4 M Sodium phosphate monobasic monohydrate / Potassium phosphate dibasic pH 6.9. Please note items from this supplier are non-returnable.