Learn More
Hampton Research QUIK SCREEN NOC5 185ML
SDP
Catalog No. 501816
Encompass
HR2-921-C5/Quik Screen #C5/Grid Screen Salt HT (F05), 185 ml - 1.4 M Sodium phosphate monobasic monohydrate / Potassium phosphate dibasic pH 7.5. Please note items from this supplier are non-returnable.
Catalog No. 50-181-6 Supplier Hampton Research Supplier No. HR2921C5
HR2-921-C5/Quik Screen #C5/Grid Screen Salt HT (F05), 185 ml - 1.4 M Sodium phosphate monobasic monohydrate / Potassium phosphate dibasic pH 7.5. Please note items from this supplier are non-returnable.