Learn More
Hampton Research QUIK SCREEN NOD1 185ML
SDP
Catalog No. 501818
Encompass
HR2-921-D1/Quik Screen #D1/Grid Screen Salt HT (F07), 185 ml - 1.8 M Sodium phosphate monobasic monohydrate / Potassium phosphate dibasic pH 5.0. Please note items from this supplier are non-returnable.
Catalog No. 50-181-8 Supplier Hampton Research Supplier No. HR2921D1
HR2-921-D1/Quik Screen #D1/Grid Screen Salt HT (F07), 185 ml - 1.8 M Sodium phosphate monobasic monohydrate / Potassium phosphate dibasic pH 5.0. Please note items from this supplier are non-returnable.