Learn More
Hampton Research QUIK SCREEN NOD2 185ML
SDP
Catalog No. 501819
Encompass
HR2-921-D2/Quik Screen #D2/Grid Screen Salt HT (F08), 185 ml - 1.8 M Sodium phosphate monobasic monohydrate / Potassium phosphate dibasic pH 5.6. Please note items from this supplier are non-returnable.
Catalog No. 50-181-9 Supplier Hampton Research Supplier No. HR2921D2
HR2-921-D2/Quik Screen #D2/Grid Screen Salt HT (F08), 185 ml - 1.8 M Sodium phosphate monobasic monohydrate / Potassium phosphate dibasic pH 5.6. Please note items from this supplier are non-returnable.