Learn More
Hampton Research QUIK SCREEN NOD3 185ML
SDP
Catalog No. 501820
Encompass
HR2-921-D3/Quik Screen #D3/Grid Screen Salt HT (F09), 185 ml - 1.8 M Sodium phosphate monobasic monohydrate / Potassium phosphate dibasic pH 6.3. Please note items from this supplier are non-returnable.
Catalog No. 50-182-0 Supplier Hampton Research Supplier No. HR2921D3
HR2-921-D3/Quik Screen #D3/Grid Screen Salt HT (F09), 185 ml - 1.8 M Sodium phosphate monobasic monohydrate / Potassium phosphate dibasic pH 6.3. Please note items from this supplier are non-returnable.