Learn More
Hampton Research QUIK SCREEN NOD4 185ML
SDP
Catalog No. 501821
Encompass
HR2-921-D4/Quik Screen #D4/Grid Screen Salt HT (F10), 185 ml - 1.8 M Sodium phosphate monobasic monohydrate / Potassium phosphate dibasic pH 6.9. Please note items from this supplier are non-returnable.
Catalog No. 50-182-1 Supplier Hampton Research Supplier No. HR2921D4
HR2-921-D4/Quik Screen #D4/Grid Screen Salt HT (F10), 185 ml - 1.8 M Sodium phosphate monobasic monohydrate / Potassium phosphate dibasic pH 6.9. Please note items from this supplier are non-returnable.