Learn More
Hampton Research QUIK SCREEN NOD5 185ML
SDP
Catalog No. 501822
Encompass
HR2-921-D5/Quik Screen #D5/Grid Screen Salt HT (F11), 185 ml - 1.8 M Sodium phosphate monobasic monohydrate / Potassium phosphate dibasic pH 7.5. Please note items from this supplier are non-returnable.
Catalog No. 50-182-2 Supplier Hampton Research Supplier No. HR2921D5
HR2-921-D5/Quik Screen #D5/Grid Screen Salt HT (F11), 185 ml - 1.8 M Sodium phosphate monobasic monohydrate / Potassium phosphate dibasic pH 7.5. Please note items from this supplier are non-returnable.