Learn More
Hampton Research QUIK SCREEN NOD6 185ML
SDP
Supplier:  Hampton Research HR2921D6
Encompass
HR2-921-D6/Quik Screen #D6/Grid Screen Salt HT (F12), 185 ml - 1.8 M Sodium phosphate monobasic monohydrate / Potassium phosphate dibasic pH 8.2. Please note items from this supplier are non-returnable.
Catalog No. 50-182-3