Learn More
Agilent Technologies QUIKCHANGE II XL10 REACTIONS
Catalog No. NC9045762
Encompass
Kit; Mutagenesis; QuikChange II XL site-directed mutagenesis kit, Contains: PfuUltra High Fidelity DNA polymerase, 10X Reaction buffer, QuikSolution reagent, dNTP mix, Dpn I restriction enzyme, QuikChange XL control plasmid and primers
Catalog No. NC9045762 Supplier Agilent Technologies Supplier No. 200521
Kit; Mutagenesis; QuikChange II XL site-directed mutagenesis kit, Contains: PfuUltra High Fidelity DNA polymerase, 10X Reaction buffer, QuikSolution reagent, dNTP mix, Dpn I restriction enzyme, QuikChange XL control plasmid and primers