Learn More
Agilent Technologies QUIKCHANGE LIGHTN SITE-DIRECT
Catalog No. NC9612995
Encompass
Contains optimized enzyme formulations that maintain the same accuracy and efficiency of previous QuikChange kits while performing the reaction at up to 50% faster.
Catalog No. NC9612995 Supplier Agilent Technologies Supplier No. 210519
Contains optimized enzyme formulations that maintain the same accuracy and efficiency of previous QuikChange kits while performing the reaction at up to 50% faster.