Learn More
Agilent Technologies QUIKCHANGE LIGHTNING MULTI
Catalog No. NC9826693
Encompass
QuikChange Lightning Multi Site-Directed Mutagenesis Kit contains optimized enzyme formulations that maintain the same accuracy and efficiency of previous QuikChange kits while performing the reaction at up to 50% faster.
Catalog No. NC9826693 Supplier Agilent Technologies Supplier No. 210514
QuikChange Lightning Multi Site-Directed Mutagenesis Kit contains optimized enzyme formulations that maintain the same accuracy and efficiency of previous QuikChange kits while performing the reaction at up to 50% faster.