Learn More
Agilent Technologies QUIKCHANGE LIGHTNING MULTI-SIT
Catalog No. NC9893312
Encompass
Academic use only. QuikChange Lightning Multi Site-Directed Mutagenesis Kit contains optimized enzyme formulations that maintain the same accuracy and efficiency of previous QuikChange kits while performing the reaction at up to 50% faster.
Catalog No. NC9893312 Supplier Agilent Technologies Supplier No. 210515
Academic use only. QuikChange Lightning Multi Site-Directed Mutagenesis Kit contains optimized enzyme formulations that maintain the same accuracy and efficiency of previous QuikChange kits while performing the reaction at up to 50% faster.