Learn More
Agilent Technologies QUIKCHANGE MULTIMUTAKIT-COMM
Catalog No. 50125159
Encompass
QuikChange Multi-Site Directed Mutagenesis kit 1 kit 30 reactions The kit offers a novel robust technology for complex multi-site mutagenesis in a single reaction Applications Randomize amino acid substitutions at key sites
Catalog No. 50-125-159 Supplier Agilent Technologies Supplier No. 200513
Add to Cart
Add to Cart
QuikChange Multi-Site Directed Mutagenesis kit 1 kit 30 reactions The kit offers a novel robust technology for complex multi-site mutagenesis in a single reaction Applications Randomize amino acid substitutions at key sites