Learn More
Agilent Technologies QUIKCHANGE MUTAGENESIS KITQUI
Supplier:  Agilent Technologies 200518
Encompass
QuikChange Site-Directed Mutagenesis Kit, Compatible with virtually all double-stranded plasmids, 150 times more accurate than PCR-based mutagenesis, Rapid 4-step procedure can be completed in 1 day
Catalog No. 50-125-163