Learn More
Agilent Technologies QUIKHYB HYBRIDIZATIONSOLUTION
Supplier:  Agilent Technologies 201220
Encompass
QuikHyb Rapid Hybridization Solution. Reduces hybridization time from 12-24 hours to 1-2 hours, Efficient and sensitive radioactive and nonradioactive detection of NAs. Applications: Southern, Northern, slot-blot and plaque lift experiments
Catalog No. 50-125-194
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.