Learn More
TARGETMOL CHEMICALS INC QUILSECONAZOLE FORMIC AC 5MG
SDP
Catalog No. 502581162
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Quilseconazole Formic acid is a selective inhibitor of fungal Cyp51 with potent activities against Cryptococcus neoformans and Cryptococcus gattii.
Catalog No. 50-258-1162 Supplier TARGETMOL CHEMICALS INC Supplier No. T16704L5MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Quilseconazole Formic acid is a selective inhibitor of fungal Cyp51 with potent activities against Cryptococcus neoformans and Cryptococcus gattii.