Learn More
Contec™ Quiltec™ I Mop Head
BIOPHARMA
Catalog No. 19168530MRK Shop All Contec Inc. Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
19-168-530MRK Non-sterile
19-168-531MRK Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-168-530MRK Supplier Contec™ Supplier No. GCQT0100MRK
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
Contec Quiltec I Mop Heads are manufactured from 100% 2-ply polyester around a rayon core so are ideal for even the highest grade of cleanroom as they generate very small numbers of particle and fibres.
  • Two-ply 100% knitted polyester over a rayon core
  • Gamma irradiated at no less than 25 kGy
  • 5 mops double bagged
  • Approx 18-20 sqm coverage
  • Certificates of conformity supplied with every batch
Specifications
Color Code White
Product Type Mop Head
Sterility Non-sterile
Autoclavable Not Autoclavable
Size 39 x 13 cm
Disposable Single-use
Quantity 5/Bag, 8 Bags/Cs.
Color-coded No
Material Quilted Polyester