Learn More
Medchemexpress LLC Quin C1 | 786706-21-6 | 98.5% | 445.51 | 25 MG
SDP
Catalog No. 5000345902
Encompass_Preferred
Quin C1 is a highly specific and potent agonist for formyl peptide receptor 2 (FPR2/ALX). Quin-C1 significantly reduces the neutrophil and lymphocyte counts in BALF, diminishes expression of TNF-α, IL-1β, KC, and TGF-β1, and decreases collagen deposition in lung tissue. Quin C1 has the potential for the research of lung injury.
  • Highly specific and potent agonist for formyl peptide receptor 2 (FPR2/ALX).
  • Significantly reduces neutrophil and lymphocyte counts in BALF.
  • Diminishes expression of TNF-α, IL-1β, KC, and TGF-β1.
  • Decreases collagen deposition in lung tissue.
  • Potential for research of lung injury.
Catalog No. 50-003-45902 Supplier Medchemexpress LLC Supplier No. 25MGHY110246
Quin C1 is a highly specific and potent agonist for formyl peptide receptor 2 (FPR2/ALX). Quin-C1 significantly reduces the neutrophil and lymphocyte counts in BALF, diminishes expression of TNF-α, IL-1β, KC, and TGF-β1, and decreases collagen deposition in lung tissue. Quin C1 has the potential for the research of lung injury.
  • Highly specific and potent agonist for formyl peptide receptor 2 (FPR2/ALX).
  • Significantly reduces neutrophil and lymphocyte counts in BALF.
  • Diminishes expression of TNF-α, IL-1β, KC, and TGF-β1.
  • Decreases collagen deposition in lung tissue.
  • Potential for research of lung injury.