Learn More
Medchemexpress LLC Quin C1 | 786706-21-6 | 98.5% | 445.51 | 500 UG
SDP
Catalog No. 5000344231
Encompass_Preferred
Quin C1 is a highly specific and potent agonist for formyl peptide receptor 2 (FPR2/ALX). It significantly reduces neutrophil and lymphocyte counts in BALF, diminishes expression of TNF-α, IL-1β, KC, and TGF-β1, and decreases collagen deposition in lung tissue.
  • Highly specific and potent agonist for formyl peptide receptor 2 (FPR2/ALX)
  • Significantly reduces neutrophil and lymphocyte counts in BALF
  • Diminishes expression of TNF-α, IL-1β, KC, and TGF-β1
  • Decreases collagen deposition in lung tissue
  • Potential for research into lung injury
Catalog No. 50-003-44231 Supplier Medchemexpress LLC Supplier No. HY11024610MG
Quin C1 is a highly specific and potent agonist for formyl peptide receptor 2 (FPR2/ALX). It significantly reduces neutrophil and lymphocyte counts in BALF, diminishes expression of TNF-α, IL-1β, KC, and TGF-β1, and decreases collagen deposition in lung tissue.
  • Highly specific and potent agonist for formyl peptide receptor 2 (FPR2/ALX)
  • Significantly reduces neutrophil and lymphocyte counts in BALF
  • Diminishes expression of TNF-α, IL-1β, KC, and TGF-β1
  • Decreases collagen deposition in lung tissue
  • Potential for research into lung injury